missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Phospholipase A2 (C. adamanteus)

Product Code. 16131830
Change view
Click to view available options
:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16131830 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16131830 Supplier Abnova™ Supplier No. P5302.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 30kDa
Name Phospholipase A2 (C. adamanteus)
Preparation Method Native protein chromatographically purified from Crotalus adamanteus Venom
Quantity 1 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name PHOSPHOLIPASE
Assays Phospholipase A2 Assay
Biological Activity ≥200 units per mg dry weight. One Unit releases one micromole of acid from soybean lecithin per minute at 25°C, pH 8.9.
Species Snake
Protein Tag None
Expression System Native protein chromatographically purified from Crotalus adamanteus Venom
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.