missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phosphodiesterase I

Product Code. 15885178 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 U
Unit Size:
100 units
This item is not returnable. View return policy

Product Code. 15885178

Brand: Thermo Scientific™ J20240.!R

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extracted from Crotalus adamanteus venom.

Phosphodiesterase I is widely used in studying nucleic acid structure and sequence. It hydrolyzes 5'-mononucleotides from 3'-hydroxy-terminated DNA and RNA. Phosphodiesterase I cleaves ADP-ribosylated proteins at the pyrophosphate linkages to yield phosphoribosyl-AMP.
  • Format: Lyophilized powder
  • Concentration: ≥20U/mg
  • Synonym: E.C. 3.1.4.1, Oligonucleate 5'-nucleotidohydrolase
  • CAS: 9025-82-5
  • MDL Number: MFCD00131883
Storage
Store at -20°C.
TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥20 U/mg
Content And Storage -20°C
Source Crotalus adamanteus venom
Quantity 100 U
Product Type Phosphodiesterase I
Form Lyophilized Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.