missing translation for 'onlineSavingsMsg'
Learn More

Phospho-XIAP (Ser87) Polyclonal Antibody, Invitrogen™

Product Code. 15565271
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15565271 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15565271 Supplier Invitrogen Supplier No. PA538349

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-XIAP (Ser87)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene XIAP
Gene Accession No. P98170, Q60989, Q9R0I6
Gene Alias API3, BIR4, BIRC4, Baculoviral IAP repeat-containing protein 4, HILP, IAP-like protein, IAP3, Inhibitor of apoptosis protein 3, X-linked IAP, X-linked inhibitor of apoptosis protein
Gene Symbols XIAP
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human XIAP around the phosphorylation site of Ser87 (K-V-SP-P-N)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11798, 331, 63879
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.