Learn More
Phospho-Vav-1 (Y160) Mouse anti-Human, Clone: 751411, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  R&D Systems MAB37861-SP
Encompass
Product Code. 15746494
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
Vav-1 Monoclonal specifically detects Vav-1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.