missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-SUFU (Ser342) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15554731
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15554731 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15554731 Supplier Invitrogen™ Supplier No. PA537794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-SUFU (Ser342) Polyclonal Antibody for Western Blot

The Hedgehog signaling pathway plays an important role in early human development. The pathway is a signaling cascade that plays a role in pattern formation and cellular proliferation during development. This gene encodes a negative regulator of the hedgehog signaling pathway. Defects in this gene are a cause of medulloblastoma. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SUFU (Ser342)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SUFU
Gene Accession No. Q9UMX1
Gene Alias OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; PRO1280; RP11-47A8.1; Su(fu); Sufu; SUFU negative regulator of hedgehog signaling; SUFUH; SUFUXL; suppressor of fused homolog; suppressor of fused homolog (Drosophila); UNQ650/PRO1280
Gene Symbols SUFU
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Serine 342 (A-P-S(p)-R-K) derived from Human Sufu.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51684
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.