missing translation for 'onlineSavingsMsg'
Learn More

Phospho-RON (Ser1394) Polyclonal Antibody, Invitrogen™

Product Code. 15585361
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15585361 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15585361 Supplier Invitrogen Supplier No. PA538441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Macrophage-stimulatin 1 receptor (MST1R/Ron), a HGF Receptor/MET-type protein kinase, mediates the biological activities of macrophage-stimulating protein (MSP), a multifunctional cytokine that regulates cell adhesion, motility, growth, and survival. The protein is a membrane-spanning, disulfide-linked heterodimer, which results from cleavage of a glycosylated precursor into 35-kD (alpha) and 150-kD (beta) subunits. Ligand binding results in tyrosine phosphorylation of the beta chain. In knockout studies, MST1R/RON (-/-) mice failed to survive past the periimplantation period. The MST1R/RON gene has been mapped to 3p21, a region of frequent deletion or mutation in small cell lung and renal carcinoma, and has been implicated in the progression of several epithelial cancers. MST1R/Ron expression has been documented in many normal human tissues. ESTs have been isolated from several tissue libraries, including normal colon, mouth, prostate, and testis and cancerous colon, prostate, stomach, and uteru.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RON (Ser1394)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MST1R
Gene Accession No. Q04912, Q62190
Gene Alias CD136, CDw136, PTK8, RON, MSP receptor, MST1R variant RON30, MST1R variant RON62, PTK8 protein tyrosine kinase 8, RON variant E2E3, c-met-related tyrosine kinase, macrophage-stimulating protein receptor, p185-Ron, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble R
Gene Symbols MST1R
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human CD136/RON around the phosphorylation site of Ser1394 (P-L-SP-E-P)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19882, 4486
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.