missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PKD2 (Ser812) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16681015
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16681015 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16681015 Supplier Invitrogen™ Supplier No. PA5104567

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of PKD2 only when phosphorylated at Ser812.

This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal dominant polycystic kidney disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PKD2 (Ser812)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PKD2
Gene Accession No. O35245, Q13563
Gene Alias A MGC138466; APKD2; Autosomal dominant polycystic kidney disease type II protein; C030034P18Rik; MGC138468; PC2; Pc-2; PKD2; PKD4; polycystic kidney disease 2; polycystic kidney disease 2 (autosomal dominant); polycystic kidney disease 2 homolog; Polycystic kidney disease 2 protein; Polycystic kidney disease 2 protein homolog; polycystin 2; polycystin 2, transient receptor potential cation channel; polycystin-2; Polycystwin; R48321; RGD1559992; transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2; TRPP1; TRPP2
Gene Symbols PKD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKD2(Accession Q13563), corresponding to amino acid residues around phosphorylated Ser812.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18764, 498328, 5311
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.