missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Myb (Ser532) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16681225
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16681225 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16681225 Supplier Invitrogen™ Supplier No. PA5104781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of c-Myb only when phosphorylated at Serine 532.

Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5'-YAAC[GT]G-3'. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Myb (Ser532)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYB
Gene Accession No. P06876, P10242
Gene Alias AI550390; Cmyb; c-myb; cmyb protein; c-myb_CDS; c-myb10A_CDS; c-myb13A_CDS; c-myb14A_CDS; c-myb8B_CDS; efg; gag-myb protein; M16449; MYB; myb proto-oncogene protein; MYB proto-oncogene, transcription factor; Mybl2; myb-related protein B; myeloblastosis oncogene; myeloblastosis oncogene-like 2; myeloblastosis proto-oncogene product; oncogene AMV; proto-oncogene c-Myb; RP1-32B1.1; transcription factor cmyb; transcriptional activator Myb; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog
Gene Symbols MYB
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYB(Accession P10242), corresponding to amino acid residues around phosphorylated Ser532.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17863, 4602
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.