missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Phospho-IkB Alpha (Ser32/36) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-IkB Alpha (Ser32/36). This antibody recognizes human antigen. The Phospho-IkB Alpha (Ser32/36) antibody has been validated for the following applications: WB, IHC, FC (Intra), ELISA analysis.
Specifications
Specifications
| Antigen | IkB Alpha (Ser32/36) |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Recombinant Monoclonal |
| Clone | 1G21 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Gene Symbols | IKBA; MAD-3; NFKBI |
| Host Species | Rabbit |
| Immunogen | Peptide |
| Purification Method | Protein A purification |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?