missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-HDAC6 (Ser22) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538498
This item is not returnable.
View return policy
Description
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
Specifications
| Phospho-HDAC6 (Ser22) | |
| Polyclonal | |
| Unconjugated | |
| HDAC6 | |
| HD6, HDA6, Histone deacetylase 6, JM21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10013, 15185 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UBN7, Q9Z2V5 | |
| HDAC6 | |
| A synthetic phosphopeptide derived from human HDAC6 around the phosphorylation site of Ser22 (P-Q-SP-P-P) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction