missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-GYS1 (Ser645) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16611245
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16611245 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16611245 Supplier Invitrogen™ Supplier No. PA5104794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Glycogen Synthase only when phosphorylated at Serine 645.

Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GYS1 (Ser645)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gys1
Gene Accession No. A2RRU1, P13807, Q9Z1E4
Gene Alias EC 2.4.1.11; glycogen [starch] synthase, muscle; glycogen synthase; glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 1, muscle; glycogen synthase 3, brain; GSY; GYS; GYS1; Gys3; MGS
Gene Symbols Gys1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GYS1(Accession P13807), corresponding to amino acid residues around phosphorylated Ser645.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14936, 2997, 690987
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.