missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-GATA3 (Ser308) Recombinant Rabbit Monoclonal Antibody (9L8O7)

Product Code. 17106523
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17106523 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17106523 Supplier Invitrogen™ Supplier No. MA535863
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Phospho-GATA3 (Ser308) Recombinant Monoclonal Antibody for ELISA, Western Blot

The genes for all 4 subunits of the T-cell antigen receptor (alpha, beta, gamma and delta) are controlled by distinct enhancers and their enhancer-binding proteins. Marine and Winoto (1991) identified a common TCR regulatory element by demonstrating binding of the enhancer-binding protein GATA3 to the enhancer elements of all 4 TCR genes. GATA3 had been shown in the chicken to be an enhancer-binding protein containing a zinc finger domain. GATA3 mRNA was demonstrated by Northern blot analysis in T cells but not in B cells or macrophages. GATA3 is abundantly expressed in the T-lymphocyte lineage and is thought to participate in T-cell receptor gene activation through binding to enhancers. Labastie et al. (1994) cloned the human gene and the 5-prime end of the mouse gene. The human gene comprises 6 exons distributed over 17 kb of DNA. Its 2 zinc fingers are encoded by 2 separate exons highly conserved with those of GATA1, but no other structural homologies between the 2 genes could be found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GATA3 (Ser308)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9L8O7
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GATA3
Gene Accession No. P23772
Gene Alias EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3
Gene Symbols GATA3
Host Species Rabbit
Immunogen A phospho specific peptide corresponding to residues surrounding S308 of human GATA3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14462
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.