missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EphA3 (Tyr602) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106128
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of EPHA3 only when phosphorylated at Tyr602.
EPHA3 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA3 encodes a protein that binds ephrin-A ligands and receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EPHA3 binds ephrin-A ligands.
Specifications
| Phospho-EphA3 (Tyr602) | |
| Polyclonal | |
| Unconjugated | |
| EPHA3 | |
| AW492086; Cek4; EK4; End3; EPH receptor A3; EPHA3; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ETK; ETK1; H TYRO4; Hek; Hek4; Human embryo kinase; human embryo kinase 1; I79_002835; Mek4; rEK4; testicular tissue protein Li 64; TYRO4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase TYRO4 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 13837, 2042, 29210 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08680, P29319, P29320 | |
| EPHA3 | |
| A synthesized peptide derived from human EPHA3(Accession P29320), corresponding to amino acid residues around phosphorylated Tyr602. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction