Learn More
Invitrogen™ Phospho-eNOS (Ser1177) Recombinant Rabbit Monoclonal Antibody (PS01-10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA549606
Description
Sequence Similarities: No Cross with Mouse and Rat. Tissue Specificity: Platelets, placenta, liver and kidney. Positive Control: HAEC serum starved then treated with 100nM Insulin for 1 hour cell lysate, human placenta tissue. Subcellular Location: Cell membrane, Membrane, caveola, Cytoplasm, cytoskeleton, Golgi apparatus. Predicted band size: 133 kDa.
Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) and endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains.
Specifications
| Phospho-eNOS (Ser1177) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P29474 | |
| NOS3 | |
| Synthetic phosphopeptide corresponding to residues surrounding Ser1177 of human eNOS. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| PS01-10 | |
| Unconjugated | |
| NOS3 | |
| 2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4846 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.