missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Claudin 5 (Tyr217) Polyclonal Antibody
Description
A suggested positive control for IHC is human brain tissue.
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
Specifications
| Antigen | Phospho-Claudin 5 (Tyr217) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CLDN5 |
| Gene Accession No. | O00501, O54942, Q9JKD6 |
| Gene Alias | AI854493; AWAL; BEC1; Brain endothelial cell clone 1 protein; claudin 5; claudin 5 (transmembrane protein deleted in velocardiofacial syndrome); claudin-5; claudin-5 protein; Cldn5; CPETRL1; lung-specific membrane protein; MBEC1; TMDVCF; TMVCF; Transmembrane protein deleted in VCFS; transmembrane protein deleted in velocardiofacial syndrome; Unknown (protein for MGC:138054) |
| Gene Symbols | CLDN5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?