missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Caspase 1 (Ser376) Polyclonal Antibody, Invitrogen™
Description
Caspases are a family of cysteine proteases that are key mediators of programmed cell death or apoptosis (1). The precursor form of all caspases is composed of a prodomain, and large and small catalytic subunits. The active forms of caspases are generated by several stimuli including ligand-receptor interactions, growth factor deprivation and inhibitors of cellular functions. All known caspases require cleavage adjacent to aspartates to liberate one large and one small subunit, which associate into a tetramer to form the active enzyme. Caspase-1/ICE (IL-1b converting enzyme) is similar to the cell death gene CED-3 of Caenorhabditilis elegans and regulates multiple proinflammatory cytokines, including interleukin-1b and interferon-gamma-inducing factor.
Specifications
Specifications
| Antigen | Phospho-Caspase 1 (Ser376) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CASP1 |
| Gene Accession No. | P29466, P29452 |
| Gene Alias | ICE, IL1BC, P45, CASP1 nirs variant 1, IL-1 beta-converting enzyme, IL1B-convertase, caspase 1, apoptosis-related cysteine peptidase interleukin 1, beta, convertase, caspase-1, interleukin 1, beta, convertase, interleukin 1-B converting enzyme |
| Gene Symbols | CASP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?