Learn More
Invitrogen™ Phospho-CaMKII alpha/beta (Thr286) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14614
Description
This antibody is predicted to react with human, mouse and Xenopus based on 100% sequence homology. This antibody is specific for the ~50 kDa alpha-CaM Kinase II subunit and the ~60 kDa beta-CaM Kinase II subunit phosphorylated at Thr286 in Western blots of rat brain lysates. Immunolabeling is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide.
Ca2+/Calmodulin-Dependent Protein Kinase II (CaM Kinase II) is a multi-functional calcium and calmodulin-dependent protein kinase that mediates cellular response to a wide variety of intercellular signals (Kennedy, 1998; Schulman and Hanson, 1993). CaM Kinase II has been shown to regulate diverse cellular functions including synaptic plasticity, neurotransmitter synthesis and release, gene expression, ion channel function, carbohydrate metabolism, cytoskeletal function, and Ca2+-homeostasis (Gleason et al., 2003; Soderling, 2000; Hudmon and Schulman, 2002). Phosphorylation of Thr286 on the kinase produces an autonomously active form of CaM Kinase II (Meng et al., 2003; Picciotto et al., 1993). Autophosphorylation of Thr305 inhibits the activity CaM Kinase II. Phosphorylation at this site appears to reduce the association of CaM Kinase II with the PSD and reduces LTP and learning (Elgersma et al., 2002).
Specifications
| Phospho-CaMKII alpha/beta (Thr286) | |
| Polyclonal | |
| Unconjugated | |
| Camk2b | |
| alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca2+/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; caM kinase II subunit alpha; CaM kinase II subunit beta; CaMK II; Camk2a; Camk2b; Camk2d; CaMKII; caMK-II subunit alpha; caMK-II subunit beta; caM-kinase II alpha chain; caM-kinase II beta chain; Ck2b; mKIAA0968; PK2CDD; PKCCD; R74975 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100341507, 100342451, 12322, 12323, 24245, 25400, 815, 816 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.25 mg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P08413, P11275, P11798, P28652, Q13554, Q9UQM7 | |
| Camk2a, Camk2b | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr286 of rat CAMK2A/B conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.