missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-beta Arrestin 1 (Ser412) Recombinant Rabbit Monoclonal Antibody (23GB3845), Invitrogen™

Description
Beta Arrestin 1 is a member of a family of proteins widely expressed but especially abundant in the central nervous system. Serving as an adaptor or scaffold molecule, beta Arrestin 1 is essential for mitogenic signaling and mediates agonist-dependent desensitization and internalization of Gprotein-coupled receptors (GPCRs, e.g., beta 2-adrenergic receptor). After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs are phosphorylated by G-protein receptor kinases (GRKs) on serine residues. Beta Arrestin 1 in the cytosol is phosphorylated by ERK1 and 2 on serine412 in a negative feedback mechanism and binds to the phosphorylated receptors at the plasma membrane. Serine 412 is then dephosphorylated and the GPCRs are internalized, leading to activation of the Ras, Raf, ERK1 and 2 signaling pathway.
Specifications
Specifications
| Antigen | Phospho-beta Arrestin 1 (Ser412) |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB3845 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ARRB1 |
| Gene Accession No. | P49407 |
| Gene Alias | 1200006I17Rik; ARB1; ARR1; ARRB1; ARRB1 protein; arrdc1; arrestin; arrestin 2; arrestin beta 1; arrestin beta-1; arrestin, beta 1; arrestin-2; AW208571; BARRES; beta-arrestin; beta-arrestin1; beta-arrestin-1; G430100A01Rik; Non-visual arrestin-2 |
| Gene Symbols | ARRB1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?