missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-B-Myb (Thr494, Thr497) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564498
This item is not returnable.
View return policy
Description
Phospho-B-Myb (Thr494, 497) Polyclonal Antibody detects endogenous levels of B-Myb only when phosphorylated at Thr494, 497.
Nucleocytoplasmic transport, a signal- and energy-dependent process, takes place through nuclear pore complexes embedded in the nuclear envelope. The import of proteins containing a nuclear localization signal requires the NLS import receptor, a heterodimer of importin alpha and beta subunits also known as karyopherins. Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. In the presence of nucleoside triphosphates and the small GTP binding protein Ran, the complex moves into the nuclear pore complex and the importin subunits dissociate. Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore. Interactions between importin beta and the FG repeats of nucleoporins are essential in translocation through the pore complex. The protein encoded by this gene is a member of the importin beta family.
Specifications
| Phospho-B-Myb (Thr494, Thr497) | |
| Polyclonal | |
| Unconjugated | |
| Mybl2 | |
| B Myb; Bmyb; B-Myb; HGNC:7548; MGC15600; MYB proto-oncogene like 2; Mybl2; myb-like protein 2; Myb-related protein B; myeloblastosis oncogene-like 2; v-myb avian myeloblastosis viral oncogene homolog-like 2; v-myb myeloblastosis viral oncogene homolog (avian)-like 2; v-myb myeloblastosis viral oncogene homolog-like 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17865, 296344, 4605 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10244, P48972 | |
| Mybl2 | |
| A synthesized peptide derived from human B-Myb (Phospho-Thr494/497). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction