missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-ASK1 (Ser967) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17762744
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17762744 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17762744 Supplier Invitrogen™ Supplier No. PA5121348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat. The target is usually found in the following locations: Cytoplasm, Endoplasmic reticulum.

MAP3K5 (ASK1) is a serine/threonine kinase that is abundantly expressed in human heart and pancreas and are activated in response to various extracellular stimuli, including cytokines, growth factors and environmental stresses. A novel MAP kinase kinase kinase (MAPKKK) was identified and designated ASK1 (for apoptosis signal-regulating kinase 1) and MAPKKK5. ASK1 activates two different subgroups of MAPKK, MKK4 and MKK6, which in turn activates c-Jun N-terminal kinase (JNK) and p38 MAP kinase, respectively. ASK1/MAPKKK5 is activated by TNFR and Fas through the interaction with members of the TRAF family and Fas-associated protein Daxx. Overexpression of ASK1 induced apoptotic cell death and a catalytically inactive form of ASK1 inhibited TNF-a-induced apoptosis. ASK1 is expressed in variety of human and mouse tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ASK1 (Ser967)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MAP3K5
Gene Accession No. Q99683
Gene Alias 7420452D20Rik; apoptosis signal regulating kinase 1; apoptosis signal-regulating kinase 1; ASK; ASK 1; ASK1; ASK-1; EC 2.7.11.25; kinase ASK1; M3K5; MAP/ERK kinase kinase 5; Map3k5; MAPK/ERK kinase kinase 5; MAPKKK5; MEK kinase 5; MEKK 5; Mekk5; mitogen activated protein kinase kinase kinase 5; mitogen-activated protein kinase kinase kinase 5; RGD1306565; RP3-325F22.4
Gene Symbols MAP3K5
Host Species Rabbit
Immunogen A phospho specific peptide corresponding to residues surrounding S967 of human MAP3K5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4217
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.