missing translation for 'onlineSavingsMsg'
Learn More

Phospho-APP (Thr668) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 17521386 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17521386 20 μL 20µL
17511386 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17521386 Supplier Proteintech Supplier No. 287881AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The Phospho-APP (Thr668) antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human Phospho-APP (Thr668). This antibody recognizes human antigen. The Phospho-APP (Thr668) antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen Phospho-APP (Thr668)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.