missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-APP (Thr668) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
The Phospho-APP (Thr668) antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human Phospho-APP (Thr668). This antibody recognizes human antigen. The Phospho-APP (Thr668) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | Phospho-APP (Thr668) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 351 |
| Target Species | Human |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?