Learn More
Phosphate HR Tablets, Lovibond™
Description
Phosphate HR Tablets, Lovibond, Quantity: 100 tablets, Physical Form: Tablets
Specifications
Specifications
| Format | Tablets |
| Quantity | 100 Tablets |
| Type | Phosphate HR Tablets |
Safety and Handling
Product Identifier
- Phosphate HR
- Danger
- Reproductive toxicity Category 1B
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contain: sodium hydrogensulphate, ammonium chloride, boric acid, ammonium meta-vanadate