Learn More
Phorate Sulfone, TRC
Product Code. 30409150
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409150 100 mg 100mg
30414983 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30409150 Supplier Toronto Research Chemicals Supplier No. TRCP353495100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2588-04-7
Molecular Formula C7 H17 O4 P S3
Molecular Weight (g/mol) 292.38
Synonym Phosphorodithioic acid, O,O-diethyl (ethylsulfonyl)methyl ester (6CI), Methanethiol, (ethylsulfonyl)-, S-ester with O,O-diethyl phosphorodithioate (8CI), O,O-Diethyl S-ethylsulfonylmethyl phosphorodithioate, O,O-Diethyl S-ethylsulfonylmethyl thionophosphate, Phorate sulfone, Phosphorodithioic acid, O,O-diethyl S-[(ethylsulfonyl)methyl] ester, Phorate Sulfone
IUPAC Name diethoxy-(ethylsulfonylmethylsulfanyl)-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)SCS(=O)(=O)CC
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C7H17O4PS3/c1-4-10-12(13,11-5-2)14-7-15(8,9)6-3/h4-7H2,1-3H3
Formula Weight 292.0027
Chemical Name or Material Phorate-sulfone