Learn More
Phenylmethyl ester N-[(3aS,4R,6S,6aR)-tetrahydro-6-hydroxy-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-yl]-Carbamic acid, TRC
Specifications
Specifications
| Chemical Name or Material | Phenylmethyl ester N-[(3aS,4R,6S,6aR)-tetrahydro-6-hydroxy-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-yl]-Carbamic acid |
| CAS | 274693-53-7 |
| Molecular Formula | C16H21NO5 |
| Quantity | 1 g |
| Synonym | Carbamic acid','[(3aS,4R,6S,6aR)-tetrahydro-6-hydroxy-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-yl]-','phenylmethyl ester (9CI), Benzyl [(3aS,4R,6S,6aR)-6-hydroxy-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl]carbamate |
| InChI Formula | InChI=1S/C16H21NO5/c1-16(2)21-13-11(8-12(18)14(13)22-16)17-15(19)20-9-10-6-4-3-5-7-10/h3-7,11-14,18H,8-9H2,1-2H3,(H,17,19)/t11-,12+,13+,14-/m1/s1 |
| SMILES | O[C@H]1C[C@@H](NC(OCC2=CC=CC=C2)=O)[C@]3([H])OC(C)(C)O[C@@]31[H] |
| IUPAC Name | benzyl N-[(3aR,4S,6R,6aS)-4-hydroxy-2,2-dimethyl-4,5,6,6a-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-6-yl]carbamate |
| Molecular Weight (g/mol) | 307.34 |
| Formula Weight | 307.142 |