Learn More
Phenylmalonic Acid Mono-5-indanyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Phenylmalonic Acid Mono-5-indanyl Ester |
| CAS | 27932-00-9 |
| Molecular Formula | C18H16O4 |
| Quantity | 100 mg |
| Synonym | 5-Indanol Hydrogen Phenylmalonate, Phenylpropanedioic Acid mono(2,3-dihydro-1H-inden-5-yl) Ester, 2-Phenylpropanedioic Acid 1-(2,3-Dihydro-1H-inden-5-yl) Ester |
| InChI Formula | InChI=1S/C18H16O4/c19-17(20)16(13-5-2-1-3-6-13)18(21)22-15-10-9-12-7-4-8-14(12)11-15/h1-3,5-6,9-11,16H,4,7-8H2,(H,19,20) |
| SMILES | O=C(C(C1=CC=CC=C1)C(OC2=CC=C3C(CCC3)=C2)=O)O |
| IUPAC Name | 3-((2,3-dihydro-1H-inden-5-yl)oxy)-3-oxo-2-phenylpropanoic acid |
| Molecular Weight (g/mol) | 296.32 |
| Formula Weight | 296.1 |