Learn More
Phenylacetyl L-Glutamine, TRC
Product Code. 30008839
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008839 25 mg 25mg
30004486 10 mg 10mg
30004015 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008839 Supplier Toronto Research Chemicals Supplier No. TRCP30650025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28047-15-6
Molecular Formula C13 H16 N2 O4
Molecular Weight (g/mol) 264.28
Synonym N-Phenylacetyl-L-glutamine, Phenylacetylglutamine, L-Glutamine, N2-(2-phenylacetyl)- (ACI), L-Glutamine, N2-(phenylacetyl)- (9CI), Glutamine, N2-(phenylacetyl)-, L- (6CI, 8CI), N2-(2-Phenylacetyl)-L-glutamine (ACI), (2S)-4-Carbamoyl-2-(2-phenylacetamido)butanoic acid, (S)-5-Amino-5-oxo-2-(2-phenylacetamido)pentanoic acid, N-Phenylacetylglutamine, N2-(Phenylacetyl)-L-glutamine, NSC 203800, Phenylacetyl-L-glutamine
IUPAC Name (2S)-5-amino-5-oxo-2-[(2-phenylacetyl)amino]pentanoic acid
SMILES NC(=O)CC[C@H](NC(=O)Cc1ccccc1)C(=O)O
Specifications
InChI Formula InChI=1S/C13H16N2O4/c14-11(16)7-6-10(13(18)19)15-12(17)8-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H2,14,16)(H,15,17)(H,18,19)/t10-/m0/s1
Quantity 25 mg
Formula Weight 264.111
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Phenylacetyl L-Glutamine
Grade HPLC