missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Benzoate, TRC

Product Code. 30011838
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011838 25 g 25g
30133830 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011838 Supplier Toronto Research Chemicals Supplier No. TRCP33506025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-99-2
Formula Weight 198.0681
InChI Formula InChI=1S/C13H10O2/c14-13(11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h1-10H
IUPAC Name phenyl benzoate
Molecular Formula C13 H10 O2
Molecular Weight (g/mol) 198.22
SMILES O=C(Oc1ccccc1)c2ccccc2
Synonym Phenyl Benzoate, Benzoic acid, phenyl ester, Phenol, benzoate (7CI), NSC 37086, Phenoxy phenyl ketone, Phenyl benzoate, Phenyl ester benzoic acid
Chemical Name or Material Phenyl Benzoate

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.