missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2-pyridyl ketoxime, 99%, Thermo Scientific™

Product Code. p-7035697
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400218 5 g 5g
15410218 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400218 Supplier Thermo Scientific Alfa Aesar Supplier No. H53421.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1826-28-4
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00006310
InChI Key WBSIGZZXFQOEJK-QXMHVHEDSA-N
Synonym phenylpyridylketoxime, phenyl pyridin-2-yl methanone oxime, phenyl 2-pyridyl ketone oxime, methanone, phenyl-2-pyridinyl-, oxime, 2-benzoylpyridiine ketoxime, phenyl 2-pyridyl methanone oxime, 2-benzoylpyridine oxime, e-phenyl-2-pyridyl ketone oxime, hydroxyimino phenyl-2-pyridylmethane, ketone, phenyl 2-pyridyl, oxime 8ci
PubChem CID 5397197
IUPAC Name (2E)-2-[nitroso(phenyl)methylidene]-1H-pyridine
SMILES O=N\C(=C1/NC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 154°C to 156°C
Quantity 5 g
UN Number UN2811
Beilstein 142387
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material Phenyl 2-pyridyl ketoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.