Learn More
Phenyl 2-pyridyl ketoxime, 99%, Thermo Scientific™
Product Code. 15410218
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410218 25 g 25g
15400218 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15410218 Supplier Thermo Scientific Alfa Aesar Supplier No. H53421.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1826-28-4
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00006310
InChI Key WBSIGZZXFQOEJK-QXMHVHEDSA-N
Synonym phenylpyridylketoxime, phenyl pyridin-2-yl methanone oxime, phenyl 2-pyridyl ketone oxime, methanone, phenyl-2-pyridinyl-, oxime, 2-benzoylpyridiine ketoxime, phenyl 2-pyridyl methanone oxime, 2-benzoylpyridine oxime, e-phenyl-2-pyridyl ketone oxime, hydroxyimino phenyl-2-pyridylmethane, ketone, phenyl 2-pyridyl, oxime 8ci
PubChem CID 5397197
IUPAC Name (2E)-2-[nitroso(phenyl)methylidene]-1H-pyridine
SMILES O=N\C(=C1/NC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 154°C to 156°C
Quantity 25 g
UN Number UN2811
Beilstein 142387
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material Phenyl 2-pyridyl ketoxime