Learn More
Phentolamine Hydrochloride, TRC
Product Code. 30426896
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426896 500 mg -
30419138 250 mg 250mg
30412870 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30426896 Supplier Toronto Research Chemicals Supplier No. TRCP302550500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 73-05-2
Molecular Formula C17 H19 N3 O . Cl H
Molecular Weight (g/mol) 317.81
Synonym Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, hydrochloride (1:1), Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, monohydrochloride (9CI), Phenol, m-[N-(2-imidazolin-2-ylmethyl)-p-toluidino]-, monohydrochloride (8CI), DNP, Phentolamine chloride, Phentolamine hydrochloride, Regitine hydrochloride, m-[N-(2-Imidazolin-2-ylmethyl)-p-toluidino]phenol hydrochloride
IUPAC Name 3-[N-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-methylanilino]phenol;hydrochloride
SMILES Cl.Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C17H19N3O.ClH/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H
Formula Weight 317.1295
Percent Purity >95%
Chemical Name or Material Phentolamine Hydrochloride
Grade HPLC