missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHD4/HIF Prolyl Hydroxylase 4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
567.00 €
Specifications
| Antigen | PHD4/HIF Prolyl Hydroxylase 4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay |
| Classification | Polyclonal |
Description
PHD4/HIF Prolyl Hydroxylase 4 Polyclonal specifically detects PHD4/HIF Prolyl Hydroxylase 4 in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy.Specifications
| PHD4/HIF Prolyl Hydroxylase 4 | |
| Western Blot 1:500-1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy | |
| Polyclonal | |
| Rabbit | |
| Cancer, Hypoxia, Lipid and Metabolism | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| EC 1.14.11.-, EGLN4, HIFPH4, HIF-PH4, HIF-prolyl hydroxylase 4, HPH-4, hypoxia-inducible factor prolyl 4-hydroxylase, Hypoxia-inducible factor prolyl hydroxylase 4, P4H with transmembrane domain, P4H-TMFLJ20262, PH-4, PH4 hypoxia inducible factor prolyl 4 hydroxylase, PHD4, proline 4-hydroxylase, prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum), Prolyl hydroxlase domain-containing 4, transmembrane prolyl 4-hydroxylase | |
| P4HTM | |
| IgG | |
| Affinity Purified | |
| Human. Does not appear to work in Mouse. Rat reactivity reported in scientific literature (PMID: 17003483). |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse (Negative) | |
| NP_808808.1 | |
| 54681 | |
| The epitope recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 502) of human PHD4/HIF Prolyl Hydroxylase 4 using the numbering given in entry NP_808808.1 (GeneID 54681). | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title