missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHD2 Monoclonal Antibody (366G/76/3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516143
This item is not returnable.
View return policy
Description
The target sequence has 83% sequence homology with mouse.
SYMPK is a nuclear protein that functions in the regulation of polyadenylation and promotes gene expression. The protein forms a high-molecular weight complex with components of the polyadenylation machinery. It is thought to serve as a scaffold for recruiting regulatory factors to the polyadenylation complex. It also participates in 3'-end maturation of histone mRNAs, which do not undergo polyadenylation. The protein also localizes to the cytoplasmic plaques of tight junctions in some cell types.This gene encodes a nuclear protein that functions in the regulation of polyadenylation and promotes gene expression. The protein forms a high-molecular weight complex with components of the polyadenylation machinery. It is thought to serve as a scaffold for recruiting regulatory factors to the polyadenylation complex. It also participates in 3'-end maturation of histone mRNAs, which do not undergo polyadenylation. The protein also localizes to the cytoplasmic plaques of tight junctions in some cell types.
Specifications
| PHD2 | |
| Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% sodium azide | |
| P59722, Q91YE3, Q9GZT9 | |
| Egln1 | |
| Peptide within residues 1-50 of human PHD2/HIF Prolyl Hydroxylase 2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 366G/76/3 | |
| Unconjugated | |
| Egln1 | |
| AI503754; C1orf12; C1ORF12 antibody; ECYT3; egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia inducible factor 1; egl-9 family hypoxia-inducible factor 1; EGLN 1 EGLN1; EGLN1; HALAH; HIF PH2; HIF prolyl hydroxylase 2; HIF-P4H 2; Hif-p4h-2; HIFPH2; HIF-PH2; HIF-prolyl hydroxylase 2; HPH2; HPH-2; Hypoxia-inducible factor prolyl hydroxylase 2; ORF13; P4H2; Phd2; PHD-2; PNAS-118; PNAS-137; Prolyl Hy; Prolyl hydroxylase domain-containing protein 2; SM20; SM-20; zinc finger MYND domain-containing protein 6; ZMYND6 | |
| Mouse | |
| Protein G | |
| RUO | |
| 112405, 308913, 54583 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction