Learn More
Fisherbrand™ pH/Pt1000 Electrode
Shop All Fisherbrand Products
Click to view available options
Connector Types:
BNC Plug
DIN Plug
Unit Size:
Each
Description
- A single probe for the measurement of 2 parameters pH and temperature
- Pt1000 sensor compatible with pH meters
- Gel electrode with anti-shock epoxy body and protected head for the field
- Laboratory electrolyte glass electrode liquid
- Supplied with 1 m cable BNC or DIN plug for pH and 2 banana plugs 4 mm for the temperature
Specifications
Specifications
| Connector Types | DIN Plug |
| Diameter (Metric) | 12 mm |
| Length (Metric) | 120 mm |
| pH Range | 0 to 14 |
| Temperature (Metric) Range | 0°C to 80°C |