Learn More
SI Analytics™ pH Glass Electrode
Shop All SI Analytics Products
Click to view available options
Connector Types:
Screw Plug Head
Unit Size:
Each
Description
- Ag / AgCl reference
- Zeropoint pH = 7.0
- N-glass
Specifications
Specifications
| Description | N-glass |
| Connector Types | Screw Plug Head |
| Material | Plastic Shaft |
| pH Range | 0 to 14 |
| Type | N 2981 |
| Membrane | Sphere |
| Temperature (Metric) Range | 0°C to 80°C |
| Length (Metric) | 120 mm |
| Diameter (Metric) | 12 mm |