missing translation for 'onlineSavingsMsg'
Learn More

PH-002, MedChemExpress

Product Code. 16444708
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444708

Brand: Medchem Express HY112798

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PH-002 is an inhibitor of apolipoprotein (apo) E4 intramolecular domain interaction in neuronal cells that could rescue impairments of mitochondrial motility and neurite outgrowth.

PH-002 is an inhibitor of apolipoprotein (apo) E4 intramolecular domain interaction in neuronal cells that could rescue impairments of mitochondrial motility and neurite outgrowth.

Specifications

Chemical Name or Material PH-002
Physical Form Solid
CAS 1311174-68-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C27H33N5O4
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 75 mg/mL (152.57 mM)
SMILES O=C(N1CCN(CC2=CC=C(NC(CC3=NN(C)C(C4=C3C=CC=C4)=O)=O)C=C2)CC1)OC(C)(C)C
Molecular Weight (g/mol) 491.58
Formula Weight 491.58
Percent Purity 98.84%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.