missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: IGVMVTASHN PEEDNGVKLV DPLGEMLAPS WEEHATCLAN AEEQDMQRVL IDISEKEAVN LQQDAFVVIG RDTRPSSEKL SQSVIDGVTV LG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.
Specifications
Specifications
| Antigen | PGM3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Pgm3 |
| Gene Accession No. | O95394 |
| Gene Alias | 2810473H05Rik; acetylglucosamine phosphomutase; Agm1; BB187688; C77933; GlcNAc-P mutase; IMD23; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Pgm3; Pgm-3; Phosphoacetylglucosamine mutase; phosphoglucomutase 3; phosphoglucomutase-3 |
| Gene Symbols | Pgm3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
