missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PFI 653

Product Code. 18744524
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18744524 50 mg 50mg
18774444 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18744524 Supplier Tocris Bioscience™ Supplier No. 7963/50
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective vanin 1 inhibitor

PFI 653 is a potent and selective vanin 1 inhibitor (IC50 values in the range 6.85 - 24.5 nM). The compound exhibits selectivity over a panel of 40 kinases and a selection of 35 other targets including enzymes, receptors and ion channels (IC50 vs biotinidase >50 μM). Following administration of a single dose of PFI 653 (2 mg/kg iv) in rats AUC is 1790 ng.h/mL, half-life is 1 h and clearance is 19 mL/min/kg. Following oral administration of PFI 653 in solution, bioavailablility is 96%. In mice with DSS-induced inflammatory bowel disease (IBD), oral adminstration of PFI 653 dose-dependently reduces vanin 1 enzyme activity and disease symptoms.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Oxa-2-azaspiro[4.5]dec-2-yl[2-[(2-pyrazinylmethyl)amino]-5-pyrimidinyl]methanone
CAS 2173134-00-2
Quantity 50 mg
Target Vanin-1/VNN1
Molecular Formula C18H22N6O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.