missing translation for 'onlineSavingsMsg'
Learn More

PF-573228, MedChemExpress

Product Code. p-200097208
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16463988

Brand: Medchem Express HY10461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-573228 is a potent and selective FAK inhibitor with IC50 of 4 nM for purified recombinant catalytic fragment of FAK.

PF-573228 is a potent and selective FAK inhibitor with IC50 of 4 nM for purified recombinant catalytic fragment of FAK.

Specifications

Chemical Name or Material PF-573228
CAS 869288-64-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H20F3N5O3S
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (50.87 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=C(NC3=NC=C(C(F)(F)F)C(NCC4=CC=CC(S(=O)(C)=O)=C4)=N3)C=C2)CC1
Molecular Weight (g/mol) 491.49
Formula Weight 491.49
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.