Learn More
PF-4800567, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
PF-4800567 is a potent and selective inhibitor of casein kinase 1ε (CK1ε), with an IC50 of 32 nM, which is greater than 20-fold selectivity over CK1δ (IC50, 711 nM).
Specifications
Specifications
| Chemical Name or Material | PF-4800567 |
| CAS | 1188296-52-7 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H18ClN5O2 |
| Quantity | 25 mg |
| Solubility Information | DMSO : 75 mg/mL (208.44 mM; Need ultrasonic) |
| SMILES | NC1=C2C(N(C3CCOCC3)N=C2COC4=CC=CC(Cl)=C4)=NC=N1 |
| Molecular Weight (g/mol) | 359.81 |
| Formula Weight | 359.81 |
| Show More |