missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-4800567, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
PF-4800567 is a potent and selective inhibitor of casein kinase 1ε (CK1ε), with an IC50 of 32 nM, which is greater than 20-fold selectivity over CK1δ (IC50, 711 nM).
Specifications
Specifications
| Chemical Name or Material | PF-4800567 |
| CAS | 1188296-52-7 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H18ClN5O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 75 mg/mL (208.44 mM; Need ultrasonic) |
| SMILES | NC1=C2C(N(C3CCOCC3)N=C2COC4=CC=CC(Cl)=C4)=NC=N1 |
| Molecular Weight (g/mol) | 359.81 |
| Formula Weight | 359.81 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction