missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 3084014 hydrobromide

Description
PF 3084014 hydrobromide is a potent γ-secretase inhibitor (IC50 values are 1.2 and 6.2 nM in whole cell and cell-free assays, respectively). Reduces Aβ in brain, CSF and plasma in mice and guinea pigs.
Specifications
Specifications
| Chemical Name or Material | (2 S)-2-[[(2 S)-6,8-Difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]amino]-N-[1-[2-[(2,2-dimethylpropyl)amino]-1,1-dimethylethyl]-1 H-imidazol-4-yl]pentanamide dihydrobromide |
| CAS | 1962925-29-6 |
| Quantity | 50 mg |
| Target | gamma-Secretase |
| Molecular Formula | C27H41F2N5O.HBr |
| Formula Weight | 651.47 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?