missing translation for 'onlineSavingsMsg'
Learn More

PF-1355, MedChemExpress

Product Code. 16493198
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493198 10 mg 10mg
16499746 50 mg 50mg
16494476 100 mg 100mg
16492666 25 mg 25mg
16415937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493198 Supplier Medchem Express Supplier No. HY100873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-1355 is a selective 2-thiouracil mechanism-based MPO inhibitor, used for treatment of vasculitic diseases.

PF-1355 is a selective 2-thiouracil mechanism-based MPO inhibitor, used for treatment of vasculitic diseases.

Specifications

Chemical Name or Material PF-1355
CAS 1435467-38-1
Color Tan
Purity Grade Notes Research
Molecular Formula C14H15N3O4S
Quantity 10 mg
Synonym PF-06281355
Solubility Information DMSO : 50 mg/mL (155.59 mM; Need ultrasonic)
SMILES O=C(N)CN(C(N1)=S)C(C2=CC(OC)=CC=C2OC)=CC1=O
Molecular Weight (g/mol) 321.35
Formula Weight 321.35
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.