Learn More
PF-06747711, MedChemExpress
Product Code. 16457397
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457397 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16457397 Supplier Medchem Express Supplier No. HY112706
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress PF-06747711 is a potent, selective, and orally active retinoic acid receptor-related orphan C2 (RORC2, also known as RORγt) inverse agonist, with an IC50 of 4.1 nM. Anti-skin inflammatory activity.
PF-06747711 is a potent, selective, and orally active retinoic acid receptor-related orphan C2 (RORC2, also known as RORγt) inverse agonist, with an IC50 of 4.1 nM. Anti-skin inflammatory activity.
Specifications
Chemical Name or Material PF-06747711
CAS 1892576-58-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H26F3N5O2
Quantity 5 mg
Solubility Information DMSO : 41.67 mg/mL (83.76 mM; Need ultrasonic)
SMILES O=C(NC1=CN=C(N(C)C=C2C3CCN(C(C(C)C)=O)CC3)C2=C1C(F)(F)F)C4=CC=CC(C#N)=C4
Molecular Weight (g/mol) 497.51
Formula Weight 497.51
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation