missing translation for 'onlineSavingsMsg'
Learn More

PF-06726304, MedChemExpress

Product Code. p-200101677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483888 10 mg 10mg
16430257 50 mg 50mg
16422896 25 mg 25mg
16416617 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483888 Supplier Medchem Express Supplier No. HY103682

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-06726304 is a potent and selective EZH2 inhibitor. PF-06726304 inhibits wild-type and Y641N mutant EZH2 with Kis of 0.7 and 3.0 nM, respectively. PF-06726304 displays robust antitumor growth activity.

PF-06726304 is a potent and selective EZH2 inhibitor. PF-06726304 inhibits wild-type and Y641N mutant EZH2 with Kis of 0.7 and 3.0 nM, respectively. PF-06726304 displays robust antitumor growth activity.

Specifications

Chemical Name or Material PF-06726304
CAS 1616287-82-1
Color White
Purity Grade Notes Research
Molecular Formula C22H21Cl2N3O3
Quantity 10 mg
Solubility Information DMSO : ≥ 15 mg/mL (33.61 mM)
SMILES O=C1N(CC2=C(C)C=C(C)NC2=O)CCC3=C1C(Cl)=C(C4=C(C)ON=C4C)C=C3Cl
Molecular Weight (g/mol) 446.33
Formula Weight 446.33
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.