Learn More
PF-06291874, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Description
PF-06291874 is a highly potent, non-peptide and orally active glucagon receptor antagonist. PF-06291874 is under the study for type 2 diabetes mellitus (T2DM).
Specifications
Specifications
| Chemical Name or Material | PF-06291874 |
| CAS | 1393124-08-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H28F3N3O4 |
| Quantity | 1 mg |
| Synonym | Glucagon receptor antagonists-4 |
| Solubility Information | DMSO : 100 mg/mL (198.61 mM; Need ultrasonic) |
| SMILES | CCC[C@H](OC1=CC(C)=C(N2C=C(C(F)(F)F)C=N2)C(C)=C1)C3=CC=C(C(NCCC(O)=O)=O)C=C3 |
| Molecular Weight (g/mol) | 503.51 |
| Show More |