Learn More
PF-06263276, MedChemExpress
Product Code. 16415967
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 16415967
Brand: Medchem Express HY101024
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress PF-06263276 (PF 6263276) is a potent and selective pan-JAK inhibitor, with IC50s of 2.2 nM, 23.1 nM, 59.9 nM and 29.7 nM for JAK1, JAK2, JAK3 and TYK2, respectively.
PF-06263276 (PF 6263276) is a potent and selective pan-JAK inhibitor, with IC50s of 2.2 nM, 23.1 nM, 59.9 nM and 29.7 nM for JAK1, JAK2, JAK3 and TYK2, respectively.
Specifications
Chemical Name or Material PF-06263276
CAS 1421502-62-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H31FN8O2
Quantity 5 mg
SMILES FC1=C(O)C=C(CC)C(C2=CC(NN=C3C4=NC5=C(CN(C(C6=NC=C(N7CCCCC7)N=C6)=O)CC5)N4)=C3C=C2)=C1
Molecular Weight (g/mol) 566.63
Formula Weight 566.63
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation