missing translation for 'onlineSavingsMsg'
Learn More

PF-05175157, MedChemExpress

Product Code. 16428157
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16428157

Brand: Medchem Express HY12942

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-05175157 is broad spectrum acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 27.0, 33.0, 23.5 and 50.4 nM for ACC1 (human), ACC2 (human), ACC1 (rat), ACC2 (rat), respectively.

PF-05175157 is broad spectrum acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 27.0, 33.0, 23.5 and 50.4 nM for ACC1 (human), ACC2 (human), ACC1 (rat), ACC2 (rat), respectively.

Specifications

Chemical Name or Material PF-05175157
Physical Form Solid
CAS 1301214-47-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H27N5O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 30 mg/mL (73.98 mM; Need ultrasonic and warming)
SMILES O=C1CC2(CCN(C(C3=CC=C4C(NC(C)=N4)=C3)=O)CC2)CC5=C1N(C(C)C)N=C5
Molecular Weight (g/mol) 405.49
Formula Weight 405.49
Percent Purity 96.13%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.