Learn More
PEX7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20614-1-AP-150UL
 View more versions of this product
Product Code. 16800175
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
This gene encodes the cytosolic receptor for the set of peroxisomal matrix enzymes targeted to the organelle by the peroxisome targeting signal 2 (PTS2). Defects in this gene cause peroxisome biogenesis disorders (PBDs), which are characterized by multiple defects in peroxisome function. There are at least 14 complementation groups for PBDs, with more than one phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene have been associated with PBD complementation group 11 (PBD-CG11) disorders, rhizomelic chondrodysplasia punctata type 1 (RCDP1), and Refsum disease (RD).
Specifications
Specifications
PEX7
Polyclonal
Unconjugated
PEX7
Peroxin 7, PEX7, PTS2 receptor, PTS2R, RCDP1, RD
Rabbit
Antigen Affinity Chromatography
RUO
18634, 308718, 5191
-20°C
Liquid
Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00628, P97865
Pex7
PEX7 Fusion Protein Ag14666
150 μL
Primary
Human, Mouse, Rat