missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Peroxidase (Horseradish)

Product Code. 16171770
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16171770

Brand: Abnova™ P5232.100mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 40kDa
Name Peroxidase (Horseradish)
Preparation Method Native protein purified from Horseradish Roots
Quantity 100 mg
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name PEROXIDASE
Assays Peroxidase Assay
Biological Activity ≥85 units per mg dry weight. One Unit decomposes 1 umole of H2O2 per minute at 25°C, pH 7.0 using aminoantipyrine and phenol.
Species Plants
Protein Tag None
Expression System Native protein purified from Horseradish Roots
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.